Assay Details
Target Gene Details
Entrez Gene ID: | 5009 |
Gene Name: | ornithine carbamoyltransferase |
Gene Aliases: |
OCTD |
Location: |
Chr.X:38352483-38421450 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTC | NM_000531.5 | NP_000522.3 | ||
| XM_017029556.1 | XP_016885045.1 | |||
| AK292757.1 | ||||
| BC074745.2 | AAH74745.1 | |||
| BC107153.1 | AAI07154.1 | |||
| BC107154.1 | AAI07155.1 | |||
| BC114496.1 | ||||
| CB158478.1 | ||||
| K02100.1 | AAA59975.1 |
More Information
Set Membership: |
|

Genomic Map