Assay Details
Target Gene Details
Entrez Gene ID: | 7102 |
Gene Name: | tetraspanin 7 |
Gene Aliases: |
A15, CCG-B7, CD231, DXS1692E, MRX58, MXS1, TALLA-1, TM4SF2, TM4SF2b |
Location: |
Chr.X:38561478-38688918 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN7 | NM_004615.3 | NP_004606.2 | ||
| AB062057.1 | BAB55824.1 | |||
| AB102665.1 | BAC81134.1 | |||
| AK293178.1 | ||||
| AK293534.1 | ||||
| AK293552.1 | ||||
| AK312343.1 | ||||
| BC018036.1 | AAH18036.1 | |||
| D10653.1 | BAA01501.1 | |||
| D29808.1 | BAA06191.1 | |||
| DB475293.1 | ||||
| L10373.1 |
More Information
Set Membership: |
|

Genomic Map