Assay Details
Target Gene Details
Entrez Gene ID: | 171483 |
Gene Name: | family with sequence similarity 9 member B |
Gene Aliases: |
TEX39B |
Location: |
Chr.X:9024232-9197176 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM9B | NM_205849.3 | NP_995321.1 | ||
| XM_011545464.2 | XP_011543766.1 | |||
| AF494344.1 | AAN07163.1 | |||
| AK128470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526672 | Chr.X:8856093 - 9048808 on Build GRCh38 | Gain |
|
| nsv515590 | Chr.X:8952896 - 9048808 on Build GRCh38 | Gain |
|
| nsv6791 | Chr.X:9029888 - 9064100 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map