Assay Details
Target Gene Details
Entrez Gene ID: | 3597 |
Gene Name: | interleukin 13 receptor subunit alpha 1 |
Gene Aliases: |
CD213A1, CT19, IL-13Ra, NR4 |
Location: |
Chr.X:118726954-118794533 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL13RA1 | NM_001560.2 | NP_001551.1 | ||
| XM_011531336.1 | XP_011529638.1 | |||
| XM_011531337.1 | XP_011529639.1 | |||
| XM_017029507.1 | XP_016884996.1 | |||
| AB209849.1 | BAD93086.1 | |||
| AK296958.1 | ||||
| AK313467.1 | ||||
| BC009960.2 | AAH09960.1 | |||
| BC015768.1 | AAH15768.1 | |||
| U81379.3 | AAD00510.3 | |||
| Y09328.1 | CAA70508.1 | |||
| Y10659.1 | CAA71669.1 |
Target Gene Details
Entrez Gene ID: | 101928242 |
Gene Name: | cell cycle control protein 50B-like |
Gene Aliases: |
- |
Location: |
Chr.X:118769312-118770213 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928242 | XM_011531419.2 | XP_011529721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv527n21 | Chr.X:118388145 - 118846744 on Build GRCh38 | Gain |
|
| esv3574403 | Chr.X:118763465 - 118773871 on Build GRCh38 | Loss |
|
| esv3559240 | Chr.X:118717538 - 118781594 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map