Assay Details
Target Gene Details
Entrez Gene ID: | 51765 |
Gene Name: | serine/threonine protein kinase 26 |
Gene Aliases: |
MASK, MST4 |
Location: |
Chr.X:132023217-132075943 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK26 | NM_001042452.1 | NP_001035917.1 | ||
| NM_001042453.1 | NP_001035918.1 | |||
| NM_016542.3 | NP_057626.2 | |||
| XM_011531349.1 | XP_011529651.1 | |||
| XM_011531350.1 | XP_011529652.1 | |||
| AB040057.2 | BAA92785.2 | |||
| AF231012.1 | AAK38484.1 | |||
| AF344882.1 | AAK29620.1 | |||
| AF344883.1 | AAK29621.1 | |||
| AK075107.1 | BAC11406.1 | |||
| AK075150.1 | BAC11435.1 | |||
| AK303583.1 | ||||
| AK314356.1 | ||||
| AK316137.1 | ||||
| BC098315.1 | AAH98315.1 | |||
| BC099843.1 | AAH99843.1 | |||
| BC103503.1 | AAI03504.1 | |||
| BT020099.1 | AAV38902.1 | |||
| DA760264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515171 | Chr.X:132028931 - 132030187 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map