Assay Details
Target Gene Details
Entrez Gene ID: | 2710 |
Gene Name: | glycerol kinase |
Gene Aliases: |
GK1, GKD |
Location: |
Chr.X:30653348-30731462 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Additional Information:
For this assay, SNP(s) [rs112324337] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map