Assay Details
Target Gene Details
Entrez Gene ID: | 27316 |
Gene Name: | RNA binding motif protein, X-linked |
Gene Aliases: |
HNRNPG, HNRPG, MRXS11, RBMXP1, RBMXRT, RNMX, hnRNP-G |
Location: |
Chr.X:136869194-136880780 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMX | NM_001164803.1 | NP_001158275.1 | ||
| AK097941.1 | 1 | 534 | ||
| AK304868.1 | ||||
| BX647131.1 |
More Information
Set Membership: |
|

Genomic Map