Assay Details
Target Gene Details
Entrez Gene ID: | 4114 |
Gene Name: | MAGE family member B3 |
Gene Aliases: |
CT3.5 |
Location: |
Chr.X:30230436-30237493 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB3 | NM_002365.4 | NP_002356.2 | ||
| XM_011545513.2 | XP_011543815.1 | |||
| XM_011545514.2 | XP_011543816.1 | |||
| AK057441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740048 | Chr.X:30231023 - 30232384 on Build GRCh38 | Deletion |
|
| esv2668620 | Chr.X:30230828 - 30234525 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138620085] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map