Assay Details
Target Gene Details
Entrez Gene ID: | 4534 |
Gene Name: | myotubularin 1 |
Gene Aliases: |
CNM, MTMX, XLMTM |
Location: |
Chr.X:150562658-150673143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTM1 | XM_005274687.2 | XP_005274744.1 | ||
| XM_011531172.1 | XP_011529474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508810 | Chr.X:150498559 - 150582211 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map