Assay Details
Target Gene Details
Entrez Gene ID: | 392490 |
Gene Name: | TPT1-like protein |
Gene Aliases: |
- |
Location: |
Chr.X:72144184-72161750 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ44635 | NM_207422.2 | NP_997305.2 | ||
| AK126598.1 | BAC86606.1 | |||
| AY572224.1 | AAS76644.1 | |||
| BC137552.1 | ||||
| BC137564.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs143758590] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map