Assay Details
Target Gene Details
Entrez Gene ID: | 159090 |
Gene Name: | family with sequence similarity 122B |
Gene Aliases: |
SPACIA2 |
Location: |
Chr.X:134769566-134797232 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM122B | NM_001166599.2 | NP_001160071.1 | ||
| NM_001166600.2 | NP_001160072.1 | |||
| NM_001170757.1 | NP_001164228.1 | |||
| XM_006724730.2 | 1 | 58 | XP_006724793.1 | |
| XM_006724732.2 | 1 | 58 | XP_006724795.1 | |
| XM_006724733.3 | 1 | 52 | XP_006724796.1 | |
| XM_011531282.1 | 1 | 58 | XP_011529584.1 | |
| XM_011531283.1 | 1 | 58 | XP_011529585.1 | |
| XM_011531284.1 | 1 | 58 | XP_011529586.1 | |
| XM_011531285.1 | 1 | 58 | XP_011529587.1 | |
| XM_011531286.1 | 1 | 58 | XP_011529588.1 | |
| XM_011531287.1 | 1 | 58 | XP_011529589.1 | |
| XM_011531288.1 | 1 | 52 | XP_011529590.1 | |
| XM_011531289.1 | 1 | 58 | XP_011529591.1 | |
| XM_011531290.1 | 1 | 58 | XP_011529592.1 | |
| XM_011531291.1 | 1 | 58 | XP_011529593.1 | |
| XM_011531292.1 | 1 | 58 | XP_011529594.1 | |
| XM_011531293.1 | 1 | 58 | XP_011529595.1 | |
| XM_011531294.1 | 1 | 58 | XP_011529596.1 | |
| XM_017029306.1 | 1 | 58 | XP_016884795.1 | |
| XM_017029307.1 | 1 | 58 | XP_016884796.1 | |
| AK124650.1 | BAC85918.1 | |||
| AK297720.1 | ||||
| BC019221.2 | AAH19221.1 | |||
| BC110846.1 | AAI10847.1 | |||
| CN303065.1 |
Target Gene Details
Entrez Gene ID: | 159091 |
Gene Name: | family with sequence similarity 122C |
Gene Aliases: |
- |
Location: |
Chr.X:134796413-134855137 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM122C | NM_001170780.1 | NP_001164251.1 | ||
| AK307219.1 |
More Information
Set Membership: |
|

Genomic Map