Assay Details
Target Gene Details
Entrez Gene ID: | 80823 |
Gene Name: | basic helix-loop-helix domain containing, class B, 9 |
Gene Aliases: |
GASP3, p60TRP |
Location: |
Chr.X:102720714-102752441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BHLHB9 | NM_001142524.1 | NP_001135996.1 | ||
| NM_001142525.1 | NP_001135997.1 | |||
| NM_001142526.1 | NP_001135998.1 | |||
| NM_001142527.1 | NP_001135999.1 | |||
| NM_001142528.1 | NP_001136000.1 | |||
| NM_030639.2 | NP_085142.1 | |||
| BC041409.2 | AAH41409.1 | |||
| DA104939.1 | ||||
| DA211842.1 | ||||
| DA397309.1 | ||||
| DA398982.1 | ||||
| DA718424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526716 | Chr.X:102582805 - 102934382 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs190566232] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map