Assay Details
Target Gene Details
Entrez Gene ID: | 105373133 |
Gene Name: | uncharacterized LOC105373133 |
Gene Aliases: |
- |
Location: |
Chr.X:13044544-13086975 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373133 | XM_011545614.2 | XP_011543916.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs137887539] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map