Assay Details
Target Gene Details
Entrez Gene ID: | 8233 |
Gene Name: | zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2 |
Gene Aliases: |
U2AF1-RS2, U2AF1L2, U2AF1RS2, URP, ZC3H22 |
Location: |
Chr.X:15789723-15825718 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRSR2 | NM_005089.3 | NP_005080.1 | ||
| XM_005274597.2 | XP_005274654.2 | |||
| XM_011545589.2 | XP_011543891.2 | |||
| XM_011545590.2 | XP_011543892.2 | |||
| XM_017029881.1 | XP_016885370.1 | |||
| XM_017029882.1 | XP_016885371.1 | |||
| XM_017029883.1 | XP_016885372.1 | |||
| AK309673.1 | ||||
| BC050451.1 | ||||
| BC113454.1 | ||||
| BC113480.1 | ||||
| D49677.1 | BAA08533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2229e212 | Chr.X:15801580 - 15813860 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map