Assay Details
Target Gene Details
Entrez Gene ID: | 392490 |
Gene Name: | TPT1-like protein |
Gene Aliases: |
- |
Location: |
Chr.X:72144184-72161750 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ44635 | NM_207422.2 | NP_997305.2 | ||
| AK126598.1 | BAC86606.1 | |||
| AY572224.1 | AAS76644.1 | |||
| BC137552.1 | ||||
| BC137564.1 |
Target Gene Details
Entrez Gene ID: | 340527 |
Gene Name: | NHS like 2 |
Gene Aliases: |
- |
Location: |
Chr.X:71911073-72153286 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHSL2 | XM_011530934.2 | 8 | 10346 | XP_011529236.1 |
| XM_017029473.1 | 8 | 9316 | XP_016884962.1 | |
| XM_017029474.1 | 8 | 5458 | XP_016884963.1 | |
| XM_017029475.1 | 4 | 8701 | XP_016884964.1 | |
| XM_017029476.1 | 4 | 4818 | XP_016884965.1 | |
| XM_017029477.1 | 4 | 9732 | XP_016884966.1 |
More Information
Set Membership: |
|

Genomic Map