Assay Details
Target Gene Details
Entrez Gene ID: | 2277 |
Gene Name: | c-fos induced growth factor |
Gene Aliases: |
VEGF-D, VEGFD |
Location: |
Chr.X:15345591-15384413 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIGF | NM_004469.4 | NP_004460.1 | ||
| AJ000185.1 | CAA03942.1 | |||
| AK223160.1 | BAD96880.1 | |||
| AK313173.1 | ||||
| BC027948.1 | AAH27948.1 | |||
| D89630.1 | BAA24264.1 | |||
| DA587496.1 | ||||
| Y12863.1 | CAA73370.1 |
Target Gene Details
Entrez Gene ID: | 100532742 |
Gene Name: | PIR-FIGF readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:15345591-15491309 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIR-FIGF | NR_037859.2 | |||
| AX740234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6813 | Chr.X:15339080 - 15383961 on Build GRCh38 | Deletion |
|
| esv3558643 | Chr.X:15353379 - 15366878 on Build GRCh38 | Deletion |
|
| nsv528343 | Chr.X:15271931 - 15385229 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148676826] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map