Assay Details
Target Gene Details
Entrez Gene ID: | 6872 |
Gene Name: | TATA-box binding protein associated factor 1 |
Gene Aliases: |
BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP |
Location: |
Chr.X:71366220-71530525 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1 | NM_001286074.1 | NP_001273003.1 | ||
| NM_004606.4 | NP_004597.2 | |||
| NM_138923.3 | NP_620278.1 | |||
| NR_104387.1 | ||||
| NR_104388.1 | ||||
| NR_104389.1 | ||||
| NR_104390.1 | ||||
| NR_104391.1 | ||||
| NR_104392.1 | ||||
| NR_104393.1 | ||||
| NR_104394.1 | ||||
| NR_104395.1 | ||||
| XM_005262295.1 | XP_005262352.1 | |||
| XM_005262296.1 | XP_005262353.1 | |||
| XM_005262297.4 | XP_005262354.1 | |||
| XM_005262300.2 | XP_005262357.1 | |||
| XM_006724682.3 | XP_006724745.1 | |||
| AB300418.1 | ||||
| BP284702.1 | ||||
| D90359.1 | BAA14374.1 | |||
| X07024.1 | CAA30073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map