Assay Details
Target Gene Details
Entrez Gene ID: | 8776 |
Gene Name: | myotubularin related protein 1 |
Gene Aliases: |
- |
Location: |
Chr.X:150692997-150765103 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR1 | NM_001306144.1 | NP_001293073.1 | ||
| NM_001306145.1 | NP_001293074.1 | |||
| NM_003828.3 | NP_003819.1 | |||
| XM_005274765.3 | XP_005274822.1 | |||
| XM_005274766.3 | XP_005274823.1 | |||
| XM_005274767.3 | XP_005274824.1 | |||
| XM_006724855.3 | XP_006724918.1 | |||
| XM_011531209.2 | XP_011529511.1 | |||
| XM_011531210.2 | XP_011529512.1 | |||
| XM_017029920.1 | XP_016885409.1 | |||
| XM_017029921.1 | XP_016885410.1 | |||
| XM_017029922.1 | XP_016885411.1 | |||
| XM_017029923.1 | XP_016885412.1 | |||
| XM_017029924.1 | XP_016885413.1 | |||
| AF057354.1 | AAD40368.1 | |||
| AJ224979.1 | CAA12271.1 | |||
| AK226109.1 | ||||
| AK295738.1 | ||||
| BC032947.1 | AAH32947.1 | |||
| DR000536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574979 | Chr.X:150708984 - 150712087 on Build GRCh38 | Loss |
|
| nsv508811 | Chr.X:150663751 - 150718401 on Build GRCh38 | Insertion |
|
| esv3577528 | Chr.X:150636374 - 150760583 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map