Assay Details
Target Gene Details
Entrez Gene ID: | 357 |
Gene Name: | shroom family member 2 |
Gene Aliases: |
APXL, HSAPXL |
Location: |
Chr.X:9786406-9949443 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHROOM2 | NM_001649.3 | NP_001640.1 | ||
| XM_005274500.4 | XP_005274557.2 | |||
| BC140866.1 | ||||
| X83543.1 | CAA58534.1 |
More Information
Set Membership: |
|

Genomic Map