Assay Details
Target Gene Details
Entrez Gene ID: | 2000 |
Gene Name: | E74 like ETS transcription factor 4 |
Gene Aliases: |
ELFR, MEF |
Location: |
Chr.X:130064920-130110713 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF4 | NM_001127197.1 | NP_001120669.1 | ||
| NM_001421.3 | NP_001412.1 | |||
| XM_005262389.3 | XP_005262446.1 | |||
| XM_011531307.2 | XP_011529609.1 | |||
| XM_011531308.2 | XP_011529610.1 | |||
| AF000670.1 | AAC17452.1 | |||
| AK314972.1 | ||||
| BC017194.2 | AAH17194.1 | |||
| U32645.1 | AAB53693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
| nsv7090 | Chr.X:130048886 - 130093759 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map