Assay Details
Target Gene Details
Entrez Gene ID: | 100533997 |
Gene Name: | MAGEA10-MAGEA5 readthrough |
Gene Aliases: |
CT1.5 |
Location: |
Chr.X:152114049-152138578 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA10-MAGEA5 | NM_001204811.2 | NP_001191740.1 | ||
| BX109673.1 | ||||
| FY212575.1 | ||||
| FY212810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520971 | Chr.X:152109627 - 152136472 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138760419] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map