Assay Details
Target Gene Details
Entrez Gene ID: | 27316 |
Gene Name: | RNA binding motif protein, X-linked |
Gene Aliases: |
HNRNPG, HNRPG, MRXS11, RBMXP1, RBMXRT, RNMX, hnRNP-G |
Location: |
Chr.X:136869194-136880780 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMX | NM_001164803.1 | NP_001158275.1 | ||
| NM_002139.3 | NP_002130.2 | |||
| NR_028476.1 | ||||
| NR_028477.1 | ||||
| AK091520.1 | ||||
| AK304573.1 | ||||
| AK304868.1 | ||||
| AK309911.1 | ||||
| AY464692.1 | AAR28036.1 | |||
| BC006550.2 | AAH06550.1 | |||
| BC007435.2 | AAH07435.1 | |||
| BM147410.1 | ||||
| BX647131.1 | ||||
| DA925805.1 | ||||
| DC337568.1 | ||||
| Z23064.1 | CAA80599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574735 | Chr.X:136878358 - 136880393 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map