Assay Details
Target Gene Details
Entrez Gene ID: | 4534 |
Gene Name: | myotubularin 1 |
Gene Aliases: |
CNM, MTMX, XLMTM |
Location: |
Chr.X:150562658-150673143 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTM1 | NM_000252.2 | NP_000243.1 | ||
| XM_005274687.2 | XP_005274744.1 | |||
| XM_011531171.1 | XP_011529473.1 | |||
| XM_011531172.1 | XP_011529474.1 | |||
| XM_011531173.2 | XP_011529475.1 | |||
| XM_017029548.1 | XP_016885037.1 | |||
| XM_017029549.1 | XP_016885038.1 | |||
| XM_017029550.1 | XP_016885039.1 | |||
| AK291710.1 | ||||
| AK297021.1 | ||||
| AK297057.1 | ||||
| AK304036.1 | ||||
| AL832715.1 | ||||
| BC030779.1 | AAH30779.1 | |||
| DC418907.1 | ||||
| U46024.1 | AAC51682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508810 | Chr.X:150498559 - 150582211 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map