Assay Details
Target Gene Details
Entrez Gene ID: | 2273 |
Gene Name: | four and a half LIM domains 1 |
Gene Aliases: |
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA |
Location: |
Chr.X:136146702-136211359 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL1 | NM_001159700.1 | NP_001153172.1 | ||
| NM_001159702.2 | NP_001153174.1 | |||
| NM_001159703.1 | NP_001153175.1 | |||
| NM_001449.4 | NP_001440.2 | |||
| NR_027621.1 | ||||
| XM_006724744.2 | XP_006724807.1 | |||
| XM_017029357.1 | XP_016884846.1 | |||
| AK091702.1 | ||||
| AK094024.1 | ||||
| AK289411.1 | ||||
| AK297124.1 | ||||
| AK300966.1 | ||||
| AK304680.1 | ||||
| BC010998.2 | AAH10998.1 | |||
| BE965488.2 | ||||
| DA497183.1 | ||||
| DC363191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7108 | Chr.X:136154921 - 136186012 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map