Assay Details
Target Gene Details
Entrez Gene ID: | 57477 |
Gene Name: | shroom family member 4 |
Gene Aliases: |
SHAP |
Location: |
Chr.X:50575534-50814167 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHROOM4 |

Genomic Map