Assay Details
Target Gene Details
Entrez Gene ID: | 6567 |
Gene Name: | solute carrier family 16 member 2 |
Gene Aliases: |
AHDS, DXS128, DXS128E, MCT 7, MCT 8, MCT7, MCT8, MRX22, XPCT |
Location: |
Chr.X:74421493-74533929 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A2 | NM_006517.4 | NP_006508.2 | ||
| AB085789.1 | BAC76827.1 | |||
| CN342861.1 | ||||
| U05315.1 | AAB60374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574055 | Chr.X:74429731 - 74431510 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map