Assay Details
Target Gene Details
Entrez Gene ID: | 9363 |
Gene Name: | RAB33A, member RAS oncogene family |
Gene Aliases: |
RabS10 |
Location: |
Chr.X:130110633-130184872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB33A | XM_017029963.1 | XP_016885452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map