Assay Details
Target Gene Details
Entrez Gene ID: | 23229 |
Gene Name: | Cdc42 guanine nucleotide exchange factor 9 |
Gene Aliases: |
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2 |
Location: |
Chr.X:63634967-63786025 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Gene Details
Entrez Gene ID: | 100874355 |
Gene Name: | ARHGEF9 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.X:63670196-63671502 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF9-IT1 | NR_046803.1 | |||
| AW138023.1 |
More Information
Set Membership: |
|

Genomic Map