Assay Details
Target Gene Details
Entrez Gene ID: | 159091 |
Gene Name: | family with sequence similarity 122C |
Gene Aliases: |
- |
Location: |
Chr.X:134796413-134855137 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM122C | NM_001170780.1 | NP_001164251.1 | ||
| XM_005262382.1 | XP_005262439.1 | |||
| XM_005262383.2 | XP_005262440.1 | |||
| XM_005262384.4 | XP_005262441.1 | |||
| XM_005262386.3 | XP_005262443.1 | |||
| XM_005262387.3 | XP_005262444.1 | |||
| XM_006724735.1 | XP_006724798.1 | |||
| XM_006724736.2 | XP_006724799.1 | |||
| XM_011531295.1 | XP_011529597.1 | |||
| XM_011531296.2 | XP_011529598.1 | |||
| XM_011531297.2 | XP_011529599.1 | |||
| XM_011531298.2 | XP_011529600.1 | |||
| XM_011531305.2 | XP_011529607.1 | |||
| XM_011531306.2 | XP_011529608.1 | |||
| XM_017029308.1 | XP_016884797.1 | |||
| XM_017029310.1 | XP_016884799.1 | |||
| AK307219.1 |

Genomic Map