Assay Details
Target Gene Details
Entrez Gene ID: | 4103 |
Gene Name: | MAGE family member A4 |
Gene Aliases: |
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B |
Location: |
Chr.X:151912889-151925170 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA4 | NM_001011548.1 | NP_001011548.1 | ||
| NM_001011549.1 | NP_001011549.1 | |||
| NM_001011550.1 | NP_001011550.1 | |||
| NM_002362.4 | NP_002353.3 | |||
| XM_005274677.2 | XP_005274734.1 | |||
| XM_005274678.3 | XP_005274735.1 | |||
| XM_005274679.3 | XP_005274736.1 | |||
| AK300179.1 | ||||
| AK313862.1 | ||||
| BC017723.1 | AAH17723.1 | |||
| BC096712.1 | AAH96712.1 | |||
| BM548926.1 | ||||
| BM562570.1 | ||||
| BX479311.1 | ||||
| CB120212.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510866 | Chr.X:151858097 - 151930089 on Build GRCh38 | Deletion |
|
| nsv953706 | Chr.X:151911729 - 151924628 on Build GRCh38 | Deletion |
|
| nsv528228 | Chr.X:151905576 - 151943206 on Build GRCh38 | Gain |
|
| esv1005324 | Chr.X:151912298 - 151922477 on Build GRCh38 | Deletion |
|
| esv22619 | Chr.X:151911966 - 151921683 on Build GRCh38 | Gain |
|
| esv2656900 | Chr.X:151910973 - 151920920 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map