Assay Details
Target Gene Details
Entrez Gene ID: | 159090 |
Gene Name: | family with sequence similarity 122B |
Gene Aliases: |
SPACIA2 |
Location: |
Chr.X:134769566-134797232 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 159090 |
Gene Name: | family with sequence similarity 122B |
Gene Aliases: |
SPACIA2 |
Location: |
Chr.X:134769566-134797232 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |