Assay Details
Target Gene Details
Entrez Gene ID: | 441525 |
Gene Name: | SPANX family member N4 |
Gene Aliases: |
CT11.9 |
Location: |
Chr.X:143025918-143038637 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPANXN4 | XM_017029543.1 | XP_016885032.1 | ||
| XM_017029544.1 | XP_016885033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664441 | Chr.X:142388728 - 143352799 on Build GRCh38 | Deletion |
|

Genomic Map