Assay Details
Target Gene Details
Entrez Gene ID: | 8905 |
Gene Name: | adaptor related protein complex 1 sigma 2 subunit |
Gene Aliases: |
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B |
Location: |
Chr.X:15825806-15855014 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1S2 | NM_001272071.1 | NP_001259000.1 | ||
| NM_003916.4 | NP_003907.3 | |||
| XM_005274614.3 | XP_005274671.1 | |||
| XM_011545599.1 | XP_011543901.1 | |||
| XM_017029925.1 | XP_016885414.1 | |||
| XM_017029926.1 | XP_016885415.1 | |||
| AB015320.1 | BAA33392.1 | |||
| AF087876.1 | AAP97176.1 | |||
| AF091077.1 | AAC72946.1 | |||
| AF251295.1 | AAG44595.1 | |||
| AK130075.1 | ||||
| AK296135.1 | ||||
| AK299921.1 | ||||
| AK302908.1 | ||||
| AK312594.1 | ||||
| BC001117.2 | AAH01117.1 | |||
| BC071867.1 | AAH71867.1 | |||
| BI462192.1 | ||||
| BT006738.1 | AAP35384.1 | |||
| BX537780.1 | CAD97839.1 |
More Information
Set Membership: |
|

Genomic Map