Assay Details
Target Gene Details
Entrez Gene ID: | 56062 |
Gene Name: | kelch like family member 4 |
Gene Aliases: |
DKELCHL, KHL4 |
Location: |
Chr.X:87517541-87670050 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL4 | NM_019117.4 | NP_061990.2 | ||
| NM_057162.2 | NP_476503.1 | |||
| AB051474.1 | ||||
| AF284765.1 | AAK49441.1 | |||
| AF284766.1 | AAK49442.1 | |||
| AK022715.1 | BAB14199.1 | |||
| AK056457.1 | ||||
| AL832514.1 | CAI46201.1 | |||
| BC140839.1 | ||||
| BC140840.1 | ||||
| BC142652.1 | ||||
| BC146677.1 | ||||
| BC161675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520176 | Chr.X:87412105 - 87607718 on Build GRCh38 | Gain |
|
| nsv520266 | Chr.X:87511338 - 87773848 on Build GRCh38 | Gain |
|
| nsv526412 | Chr.X:87210687 - 87747441 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67929085] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map