Assay Details
Target Gene Details
Entrez Gene ID: | 54830 |
Gene Name: | nucleoporin 62 C-terminal like |
Gene Aliases: |
- |
Location: |
Chr.X:107123427-107206440 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP62CL | NM_017681.2 | NP_060151.2 | ||
| NR_033676.1 | ||||
| AK000137.1 | BAA90968.1 | |||
| BC016327.1 | AAH16327.1 | |||
| BC017799.1 | AAH17799.1 | |||
| DB448067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519484 | Chr.X:106978260 - 107225478 on Build GRCh38 | Gain |
|
| esv2740376 | Chr.X:107010095 - 107327489 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map