Assay Details
Target Gene Details
Entrez Gene ID: | 260293 |
Gene Name: | cytochrome P450 family 4 subfamily X member 1 |
Gene Aliases: |
CYPIVX1 |
Location: |
Chr.1:46961364-47055432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4X1 | NM_001320289.1 | NP_001307218.1 | ||
| NM_001320290.1 | NP_001307219.1 | |||
| AK091806.1 | BAC03751.1 | |||
| AK131355.1 | BAD18508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546167 | Chr.1:47021913 - 47058592 on Build GRCh38 | Gain |
|
| nsv546166 | Chr.1:46942067 - 47029933 on Build GRCh38 | Loss |
|
| esv2763300 | Chr.1:46892860 - 47163102 on Build GRCh38 | Gain |
|
| esv3585949 | Chr.1:47009618 - 47041416 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map