Assay Details
Target Gene Details
Entrez Gene ID: | 284611 |
Gene Name: | family with sequence similarity 102 member B |
Gene Aliases: |
SYM-3B |
Location: |
Chr.1:108560349-108639327 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM102B | NM_001010883.2 | NP_001010883.2 | ||
| BC127898.1 | ||||
| BC140863.1 | ||||
| BC140865.1 | ||||
| BC150530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1009643 | Chr.1:108575281 - 108581168 on Build GRCh38 | Deletion |
|

Genomic Map