Assay Details
Target Gene Details
Entrez Gene ID: | 1382 |
Gene Name: | cellular retinoic acid binding protein 2 |
Gene Aliases: |
CRABP-II, RBP6 |
Location: |
Chr.1:156699606-156712900 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRABP2 | XM_017000343.1 | 4 | 5022 | XP_016855832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952216 | Chr.1:156704509 - 156718008 on Build GRCh38 | Deletion |
|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map