Assay Details
Target Gene Details
Entrez Gene ID: | 149297 |
Gene Name: | family with sequence similarity 78 member B |
Gene Aliases: |
- |
Location: |
Chr.1:166055918-166166755 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM78B | NM_001320302.1 | 2 | 3740 | NP_001307231.1 |
| NR_135199.1 | 3 | 4672 | ||
| XM_017000417.1 | 3 | 8878 | XP_016855906.1 | |
| XM_017000418.1 | 3 | 4220 | XP_016855907.1 | |
| AB593134.1 | 2 | 3740 | ||
| AK055347.1 | 1 | 1779 | BAB70907.1 |

Genomic Map