Assay Details
Target Gene Details
Entrez Gene ID: | 5567 |
Gene Name: | protein kinase cAMP-activated catalytic subunit beta |
Gene Aliases: |
PKA C-beta, PKACB |
Location: |
Chr.1:84077975-84238498 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKACB | NM_002731.3 | NP_002722.1 | ||
| NM_207578.2 | NP_997461.1 | |||
| XM_017001713.1 | XP_016857202.1 | |||
| AB451239.1 | ||||
| AB451364.1 | ||||
| AK074904.1 | 1 | 1605 | ||
| AK314550.1 | ||||
| BC016285.1 | AAH16285.1 | |||
| BC035058.1 | AAH35058.1 | |||
| HY160843.1 | ||||
| M34181.1 | AAA60170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546689 | Chr.1:84065434 - 84205133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map