Assay Details
Target Gene Details
Entrez Gene ID: | 8704 |
Gene Name: | beta-1,4-galactosyltransferase 2 |
Gene Aliases: |
B4Gal-T2, B4Gal-T3, beta4Gal-T2 |
Location: |
Chr.1:43979202-43991171 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT2 | NM_001005417.2 | NP_001005417.1 | ||
| NM_003780.4 | NP_003771.1 | |||
| NM_030587.2 | NP_085076.2 | |||
| XM_017002716.1 | XP_016858205.1 | |||
| XM_017002717.1 | XP_016858206.1 | |||
| AB024434.1 | BAA75819.1 | |||
| AF038660.1 | AAC39733.1 | |||
| AK074533.1 | ||||
| AK095873.1 | ||||
| AK293418.1 | ||||
| AL137647.1 | CAB70857.1 | |||
| BC096821.1 | AAH96821.1 | |||
| Y12510.1 | CAA73112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv732 | Chr.1:43976690 - 43992799 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map