Assay Details
Target Gene Details
Entrez Gene ID: | 727684 |
Gene Name: | chromosome 1 open reading frame 195 |
Gene Aliases: |
- |
Location: |
Chr.1:15164069-15171921 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf195 | NM_001278502.1 | NP_001265431.1 | ||
| XM_017002222.1 | XP_016857711.1 | |||
| BX095243.1 |
Target Gene Details
Entrez Gene ID: | 55092 |
Gene Name: | transmembrane protein 51 |
Gene Aliases: |
C1orf72 |
Location: |
Chr.1:15152418-15220480 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM51 | NM_001136216.1 | NP_001129688.1 | ||
| NM_001136217.1 | NP_001129689.1 | |||
| NM_001136218.1 | NP_001129690.1 | |||
| NM_001319665.1 | NP_001306594.1 | |||
| NM_018022.2 | NP_060492.1 | |||
| NR_135082.1 | ||||
| XM_011541676.1 | XP_011539978.1 | |||
| XM_017001590.1 | XP_016857079.1 | |||
| AK001061.1 | BAA91486.1 | |||
| AK098467.1 | ||||
| AK129922.1 | ||||
| AK292184.1 | ||||
| AL525184.3 | ||||
| BC000593.2 | AAH00593.1 | |||
| BC005177.2 | AAH05177.1 | |||
| BP275401.1 | ||||
| BQ416935.1 | ||||
| DC295166.1 | ||||
| DC345032.1 | ||||
| HY053313.1 | ||||
| HY144080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833425 | Chr.1:15083330 - 15291728 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map