Assay Details
Target Gene Details
Entrez Gene ID: | 55791 |
Gene Name: | ligand dependent nuclear receptor interacting factor 1 |
Gene Aliases: |
C1orf103, RIF1 |
Location: |
Chr.1:110877252-110964203 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIF1 | XM_017001769.1 | XP_016857258.1 |
More Information
Set Membership: |
|

Genomic Map