Assay Details
Target Gene Details
Entrez Gene ID: | 100750325 |
Gene Name: | RCAN3 antisense |
Gene Aliases: |
- |
Location: |
Chr.1:24496333-24502360 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN3AS | NR_045407.1 | |||
| NR_045408.1 | ||||
| NR_045409.1 | ||||
| HQ317448.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72882493] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map