Assay Details
Target Gene Details
Entrez Gene ID: | 477 |
Gene Name: | ATPase Na+/K+ transporting subunit alpha 2 |
Gene Aliases: |
FHM2, MHP2 |
Location: |
Chr.1:160115730-160143591 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1A2 | NM_000702.3 | NP_000693.1 | ||
| AB018321.2 | ||||
| AK091617.1 | ||||
| AK124581.1 | ||||
| AK295048.1 | ||||
| AK314296.1 | ||||
| AL831991.1 | ||||
| AL831997.1 | ||||
| BC052271.1 | AAH52271.2 | |||
| KU177927.1 | ||||
| KU177928.1 |
More Information
Set Membership: |
|

Genomic Map