Assay Details
Target Gene Details
Entrez Gene ID: | 54879 |
Gene Name: | suppression of tumorigenicity 7 like |
Gene Aliases: |
FAM4B, ST7R, STLR |
Location: |
Chr.1:112517444-112620838 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831081 | Chr.1:112392298 - 112580018 on Build GRCh38 | Loss |
|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
| esv3578114 | Chr.1:112542575 - 112546226 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map