Assay Details
Target Gene Details
Entrez Gene ID: | 100873949 |
Gene Name: | IPO9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:201688256-201829559 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO9-AS1 | NR_046696.1 |
Target Gene Details
Entrez Gene ID: | 89796 |
Gene Name: | neuron navigator 1 |
Gene Aliases: |
POMFIL3, STEERIN1, UNC53H1 |
Location: |
Chr.1:201648322-201826974 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAV1 | NM_020443.4 | NP_065176.3 | ||
| XM_011510097.1 | XP_011508399.1 | |||
| XM_011510098.1 | XP_011508400.1 | |||
| XM_011510099.1 | XP_011508401.1 | |||
| XM_011510100.1 | XP_011508402.1 | |||
| XM_011510101.1 | XP_011508403.1 | |||
| XM_011510102.1 | XP_011508404.1 | |||
| XM_017002751.1 | XP_016858240.1 | |||
| XM_017002753.1 | XP_016858242.1 | |||
| XM_017002754.1 | XP_016858243.1 | |||
| AB032977.3 | BAA86465.3 | |||
| AJ488101.1 | CAD32470.1 | |||
| AY043013.1 | AAL05591.1 | |||
| BC117683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv136e212 | Chr.1:201707431 - 201721391 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map