Assay Details
Target Gene Details
Entrez Gene ID: | 6905 |
Gene Name: | tubulin folding cofactor E |
Gene Aliases: |
HRD, KCS, KCS1, pac2 |
Location: |
Chr.1:235367360-235448968 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCE | NM_001079515.2 | NP_001072983.1 | ||
| NM_001287801.1 | NP_001274730.1 | |||
| NM_001287802.1 | NP_001274731.1 | |||
| NM_003193.4 | NP_003184.1 | |||
| AK093397.1 | ||||
| AK292287.1 | ||||
| AK296185.1 | ||||
| AK298050.1 | ||||
| BC008654.1 | AAH08654.1 | |||
| BT007086.1 | AAP35749.1 | |||
| DA418943.1 | ||||
| U61232.1 | AAB17538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013571 | Chr.1:235210731 - 235587805 on Build GRCh38 | Gain |
|
| dgv30n21 | Chr.1:235198002 - 235605078 on Build GRCh38 | Gain |
|
| nsv509668 | Chr.1:235406099 - 235430910 on Build GRCh38 | Insertion |
|
| nsv4954 | Chr.1:235408845 - 235432924 on Build GRCh38 | Insertion |
|
| esv3575794 | Chr.1:235370375 - 235549222 on Build GRCh38 | Gain |
|
| nsv549392 | Chr.1:235375705 - 235548388 on Build GRCh38 | Gain |
|
| nsv1011288 | Chr.1:235376656 - 235666302 on Build GRCh38 | Gain |
|
| nsv468406 | Chr.1:235395391 - 235463092 on Build GRCh38 | Gain |
|
| dgv586n100 | Chr.1:235367709 - 235547476 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77452241] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map