Assay Details
Target Gene Details
Entrez Gene ID: | 1573 |
Gene Name: | cytochrome P450 family 2 subfamily J member 2 |
Gene Aliases: |
CPJ2, CYPIIJ2 |
Location: |
Chr.1:59893308-59926798 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2J2 | NM_000775.3 | NP_000766.2 | ||
| NR_134981.1 | ||||
| NR_134982.1 | ||||
| AB080265.1 | BAB85489.1 | |||
| AK055088.1 | ||||
| AK315387.1 | ||||
| BC032594.1 | AAH32594.1 | |||
| CD013972.1 | ||||
| CD013973.1 | ||||
| U37143.2 | AAC50370.2 |
More Information
Set Membership: |
|

Genomic Map