Assay Details
Target Gene Details
Entrez Gene ID: | 8565 |
Gene Name: | tyrosyl-tRNA synthetase |
Gene Aliases: |
CMTDIC, TYRRS, YRS, YTS |
Location: |
Chr.1:32775237-32818032 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YARS | NM_003680.3 | NP_003671.1 | ||
| XM_011542347.1 | XP_011540649.1 | |||
| XM_011542348.1 | XP_011540650.1 | |||
| XM_017002651.1 | XP_016858140.1 | |||
| AK026535.1 | ||||
| AK125213.1 | ||||
| AK126444.1 | ||||
| AK127182.1 | ||||
| AK223608.1 | BAD97328.1 | |||
| BC001933.1 | AAH01933.1 | |||
| BC004151.2 | AAH04151.1 | |||
| BC016689.1 | AAH16689.1 | |||
| KU178431.1 | ||||
| KU178432.1 | ||||
| U89436.1 | AAB88409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585677 | Chr.1:32776270 - 32778868 on Build GRCh38 | Loss |
|
| nsv322 | Chr.1:32758359 - 32781995 on Build GRCh38 | Deletion |
|
| nsv509158 | Chr.1:32691713 - 32783820 on Build GRCh38 | Insertion |
|
| nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map